Assay Details
Target Gene Details
Entrez Gene ID: | 7709 |
Gene Name: | zinc finger and BTB domain containing 17 |
Gene Aliases: |
MIZ-1, ZNF151, ZNF60, pHZ-67 |
Location: |
Chr.1:15941869-15976132 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB17 | NM_001242884.1 | 15 | 2475 | NP_001229813.1 |
| NM_001287603.1 | 16 | 2681 | NP_001274532.1 | |
| NM_001287604.1 | 16 | 2656 | NP_001274533.1 | |
| NM_001324137.1 | 15 | 2453 | NP_001311066.1 | |
| NM_001324138.1 | 15 | 2471 | NP_001311067.1 | |
| NM_003443.2 | 16 | 2660 | NP_003434.2 | |
| XM_005245986.1 | 15 | 2492 | XP_005246043.1 | |
| XM_005245987.1 | 16 | 2622 | XP_005246044.1 | |
| XM_011542085.2 | 15 | 2467 | XP_011540387.1 | |
| XM_011542086.1 | 16 | 2648 | XP_011540388.1 | |
| XM_011542087.2 | 15 | 2489 | XP_011540389.1 | |
| XM_011542088.1 | 16 | 2627 | XP_011540390.1 | |
| XM_017002242.1 | 14 | 2462 | XP_016857731.1 | |
| AK223618.1 | 16 | 2637 | BAD97338.1 | |
| AK226168.1 | 16 | 2600 | ||
| AK301896.1 | 15 | 2422 | ||
| AK303823.1 | 15 | 2366 | ||
| AK315201.1 | ||||
| BC126163.1 | 15 | 2523 | ||
| BC143965.1 | 15 | 2544 | ||
| BQ575263.1 | 1 | 108 | ||
| BU634371.1 | 1 | 108 | ||
| U20647.1 | 4 | 684 | AAC50256.1 | |
| Y09723.1 | 16 | 2547 | CAA70889.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833647 | Chr.1:15794042 - 15952334 on Build GRCh38 | Loss |
|
| nsv954843 | Chr.1:15931306 - 15953205 on Build GRCh38 | Deletion |
|
| nsv508936 | Chr.1:15915852 - 16181025 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map