Assay Details
Target Gene Details
Entrez Gene ID: | 93436 |
Gene Name: | armadillo repeat containing 6 |
Gene Aliases: |
R30923_1 |
Location: |
Chr.19:19033578-19058178 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARMC6 | NM_001199196.1 | 5 | 910 | NP_001186125.1 |
| NM_033415.3 | 4 | 802 | NP_219483.1 | |
| XM_005260157.3 | 4 | 853 | XP_005260214.1 | |
| XM_017027484.1 | 4 | 777 | XP_016882973.1 | |
| XM_017027485.1 | 6 | 1034 | XP_016882974.1 | |
| AK295488.1 | 4 | 576 | ||
| AK303914.1 | 2 | 490 | ||
| BC067125.1 | 5 | 679 | AAH67125.1 | |
| BX648486.1 | 4 | 697 | ||
| CR749678.1 | 1 | 142 | CAH18469.1 | |
| Z36779.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952424 | Chr.19:19033592 - 19069091 on Build GRCh38 | Deletion |
|
| nsv833778 | Chr.19:18901860 - 19090678 on Build GRCh38 | Loss |
|
| nsv1151217 | Chr.19:18627369 - 19284914 on Build GRCh38 | Deletion |
|
| nsv1059305 | Chr.19:19020501 - 19099190 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map