Assay Details
Target Gene Details
Entrez Gene ID: | 8521 |
Gene Name: | glial cells missing homolog 1 |
Gene Aliases: |
GCMA, hGCMa |
Location: |
Chr.6:53125644-53262803 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCM1 | NM_003643.3 | 5 | 731 | NP_003634.2 |
| XM_017011390.1 | 9 | 2113 | XP_016866879.1 | |
| AB026493.1 | 4 | 602 | BAA77250.2 | |
| AB041714.1 | 4 | 522 | BAA94757.1 | |
| BC096288.1 | 5 | 689 | AAH96288.1 | |
| D88613.1 | 5 | 734 | BAA13651.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76623467] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map