Assay Details
Target Gene Details
Entrez Gene ID: | 83694 |
Gene Name: | ribosomal protein S6 kinase like 1 |
Gene Aliases: |
RSKL2 |
Location: |
Chr.14:74903954-74923396 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS6KL1 | NM_031464.4 | 11 | 2691 | NP_113652.2 |
| XM_011537206.2 | XP_011535508.1 | |||
| XM_011537207.2 | XP_011535509.1 | |||
| XM_011537211.2 | XP_011535513.1 | |||
| XM_011537213.2 | XP_011535515.1 | |||
| XM_017021680.1 | XP_016877169.1 | |||
| XM_017021681.1 | XP_016877170.1 | |||
| XM_017021682.1 | XP_016877171.1 | |||
| XM_017021683.1 | XP_016877172.1 | |||
| XM_017021684.1 | XP_016877173.1 | |||
| XM_017021685.1 | 13 | 2129 | XP_016877174.1 | |
| XM_017021686.1 | 11 | 2668 | XP_016877175.1 | |
| XM_017021688.1 | XP_016877177.1 | |||
| XM_017021689.1 | 10 | 1685 | XP_016877178.1 | |
| XM_017021690.1 | 9 | 2043 | XP_016877179.1 | |
| XM_017021691.1 | XP_016877180.1 | |||
| XM_017021692.1 | 9 | 1595 | XP_016877181.1 | |
| AK093053.1 | 4 | 1343 | ||
| AK131532.1 | 3 | 2725 | ||
| AK226137.1 | 9 | 2952 | ||
| AL390152.1 | 3 | 432 | CAH10661.1 | |
| BC004540.2 | AAH04540.1 |
More Information
Set Membership: |
|

Genomic Map