Assay Details
Target Gene Details
Entrez Gene ID: | 6654 |
Gene Name: | SOS Ras/Rac guanine nucleotide exchange factor 1 |
Gene Aliases: |
GF1, GGF1, GINGF, HGF, NS4 |
Location: |
Chr.2:38981549-39124959 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOS1 | NM_005633.3 | 23 | 3927 | NP_005624.2 |
| XM_005264515.4 | 22 | 4111 | XP_005264572.1 | |
| XM_011533062.2 | 23 | 4095 | XP_011531364.1 | |
| XM_011533064.2 | 23 | 3835 | XP_011531366.1 | |
| XM_011533066.2 | 16 | 2836 | XP_011531368.1 | |
| AB209140.1 | 15 | 2853 | BAD92377.1 | |
| AK290228.1 | 24 | 3973 | ||
| L13857.1 | 23 | 3886 | AAA35913.1 |
More Information
Set Membership: |
|

Genomic Map