Assay Details
Target Gene Details
Entrez Gene ID: | 384 |
Gene Name: | arginase 2 |
Gene Aliases: |
- |
Location: |
Chr.14:67619862-67651720 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARG2 | NM_001172.3 | NP_001163.1 | ||
| AK312484.1 | ||||
| AY074489.1 | AAL71548.1 | |||
| BC001350.1 | AAH01350.1 | |||
| BC008464.1 | AAH08464.1 | |||
| BC029050.1 | AAH29050.1 | |||
| CR536550.1 | CAG38787.1 | |||
| D86724.1 | BAA13158.1 | |||
| U75667.1 | AAB39855.1 | |||
| U82256.1 | AAC51664.1 |
Target Gene Details
Entrez Gene ID: | 10243 |
Gene Name: | gephyrin |
Gene Aliases: |
GEPH, GPH, GPHRYN, HKPX1, MOCODC |
Location: |
Chr.14:66507407-67735831 on Build GRCh38 |
Assay Gene Location: | Within Intron 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPHN | XM_017020925.1 | XP_016876414.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470651 | Chr.14:67266144 - 67646576 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map