Assay Details
Target Gene Details
Entrez Gene ID: | 64979 |
Gene Name: | mitochondrial ribosomal protein L36 |
Gene Aliases: |
BRIP1, L36mt, MRP-L36, PRPL36, RPMJ |
Location: |
Chr.5:1798385-1801432 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL36 | NM_032479.3 | NP_115868.1 | ||
| XM_011514079.2 | XP_011512381.1 | |||
| XM_011514080.2 | XP_011512382.1 | |||
| XM_017009750.1 | XP_016865239.1 | |||
| XM_017009751.1 | XP_016865240.1 | |||
| XM_017009752.1 | XP_016865241.1 | |||
| AB049654.1 | BAB40859.1 | |||
| AF151109.1 | AAF04788.1 | |||
| AF155653.1 | AAF67010.1 | |||
| AK312001.1 | ||||
| BC020642.1 | AAH20642.1 | |||
| BC104652.1 | AAI04653.1 | |||
| BG185418.1 | ||||
| DW465815.1 | ||||
| EF036487.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3603817 | Chr.5:1638442 - 1833414 on Build GRCh38 | Gain |
|
| nsv1033147 | Chr.5:1717188 - 1815707 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map