Assay Details
Target Gene Details
Entrez Gene ID: | 7350 |
Gene Name: | uncoupling protein 1 |
Gene Aliases: |
SLC25A7, UCP |
Location: |
Chr.4:140555770-140568914 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UCP1 | NM_021833.4 | 5 | 826 | NP_068605.1 |
| XM_005263206.3 | 5 | 932 | XP_005263263.1 | |
| XM_011532228.2 | 5 | 935 | XP_011530530.1 | |
| BC069556.1 | 5 | 826 | AAH69556.1 | |
| BC096736.1 | 5 | 823 | AAH96736.1 | |
| BC098168.1 | 5 | 826 | AAH98168.1 | |
| BC098258.1 | 5 | 826 | AAH98258.1 | |
| BC098352.1 | 5 | 823 | AAH98352.1 | |
| U28480.1 | 5 | 750 | AAA85271.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830088 | Chr.4:140436956 - 140603794 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map