Assay Details
Target Gene Details
Entrez Gene ID: | 84889 |
Gene Name: | solute carrier family 7 member 3 |
Gene Aliases: |
ATRC3, CAT-3, CAT3 |
Location: |
Chr.X:70925579-70931125 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A3 | NM_001048164.2 | 12 | 1987 | NP_001041629.1 |
| NM_032803.5 | 12 | 2011 | NP_116192.4 | |
| XM_017029912.1 | 11 | 2502 | XP_016885401.1 | |
| AF320612.1 | 12 | 1962 | AAL37184.1 | |
| AK027447.1 | 10 | 1333 | BAB55118.1 | |
| AK074865.1 | 12 | 1982 | BAC11253.1 | |
| AK075014.1 | 12 | 1987 | BAC11353.1 | |
| BC033816.1 | 12 | 1966 | AAH33816.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33082 | Chr.X:70420685 - 71888183 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map