Assay Details
Target Gene Details
Entrez Gene ID: | 420 |
Gene Name: | ADP-ribosyltransferase 4 (Dombrock blood group) |
Gene Aliases: |
ARTC4, CD297, DO, DOK1 |
Location: |
Chr.12:14829311-14843479 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ART4 | NM_021071.2 | 1 | 394 | NP_066549.2 |
| AF290204.1 | 1 | 127 | AAG17845.1 | |
| AK291662.1 | 1 | 410 | ||
| AY829431.1 | AAX39378.1 | |||
| AY829432.1 | AAX39379.1 | |||
| AY829433.1 | AAX39380.1 | |||
| AY829434.1 | 1 | 363 | AAX39381.1 | |
| AY829437.1 | AAX39384.1 | |||
| BC074727.2 | 1 | 64 | AAH74727.1 | |
| CR746266.1 | 1 | 394 |
More Information
Set Membership: |
|

Genomic Map