Assay Details
Target Gene Details
Entrez Gene ID: | 27043 |
Gene Name: | proline, glutamate and leucine rich protein 1 |
Gene Aliases: |
MNAR, P160 |
Location: |
Chr.17:4671384-4704337 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PELP1 | NM_001278241.1 | 17 | 2667 | NP_001265170.1 |
| NM_014389.2 | 16 | 2585 | NP_055204.3 | |
| AF547989.1 | 16 | 2359 | AAN41255.1 | |
| AK226056.1 | 16 | 2365 | ||
| AK291163.1 | 16 | 2385 | ||
| AK293838.1 | 17 | 2454 | ||
| AK299084.1 | 16 | 2573 | ||
| AY882602.1 | 16 | 2152 | AAW80659.1 | |
| BC002875.2 | 6 | 1200 | AAH02875.2 | |
| BC010457.2 | 15 | 2113 | AAH10457.2 | |
| BC069058.1 | 16 | 2359 | AAH69058.1 | |
| U88153.2 | 15 | 2794 | AAC17708.2 | |
| U88154.1 | 15 | 2010 | AAC17709.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
| nsv833346 | Chr.17:4646256 - 4860871 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116554252] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map