Assay Details
Target Gene Details
Entrez Gene ID: | 5307 |
Gene Name: | paired like homeodomain 1 |
Gene Aliases: |
BFT, CCF, LBNBG, POTX, PTX1 |
Location: |
Chr.5:135027734-135034274 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PITX1 | NM_002653.4 | 3 | 2314 | NP_002644.4 |
| U70370.1 | AAC51126.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv823239 | Chr.5:135019535 - 135071110 on Build GRCh38 | Gain |
|
| nsv830495 | Chr.5:135024595 - 135204207 on Build GRCh38 | Loss |
|
| nsv823243 | Chr.5:135027426 - 135033388 on Build GRCh38 | Gain |
|
| nsv823240 | Chr.5:135020631 - 135043367 on Build GRCh38 | Gain |
|
| dgv969n67 | Chr.5:135018061 - 135059172 on Build GRCh38 | Gain |
|
| nsv462458 | Chr.5:135012266 - 135070292 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map