Assay Details
Target Gene Details
Entrez Gene ID: | 389840 |
Gene Name: | mitogen-activated protein kinase kinase kinase 15 |
Gene Aliases: |
ASK3, bA723P2.3 |
Location: |
Chr.X:19360058-19515459 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K15 | NM_001001671.3 | 23 | 3204 | NP_001001671.3 |
| XM_011545507.2 | 24 | 3431 | XP_011543809.2 | |
| XM_011545508.2 | 24 | 3344 | XP_011543810.2 | |
| XM_011545510.2 | 17 | 3944 | XP_011543812.1 | |
| XM_011545511.2 | 14 | 2455 | XP_011543813.1 | |
| AK122632.1 | 2 | 622 | ||
| AK131412.1 | 20 | 2532 | BAD18559.1 | |
| AK131477.1 | 24 | 3264 | BAD18622.1 |
More Information
Set Membership: |
|

Genomic Map