Assay Details
Target Gene Details
Entrez Gene ID: | 116028 |
Gene Name: | RecQ mediated genome instability 2 |
Gene Aliases: |
BLAP18, C16orf75 |
Location: |
Chr.16:11345438-11351763 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RMI2 | NM_152308.2 | 2 | 690 | NP_689521.1 |
| NR_130754.1 | 2 | 467 | ||
| AK123764.1 | 2 | 679 | ||
| BC013040.2 | 2 | 658 | AAH13040.2 | |
| BC022427.1 | 2 | 674 | AAH22427.1 | |
| BC031016.1 | 2 | 678 | AAH31016.1 | |
| BC039361.1 | 5 | 1112 | AAH39361.1 | |
| CN311350.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2728n100 | Chr.16:11333115 - 11449038 on Build GRCh38 | Gain |
|
| nsv1040428 | Chr.16:10145408 - 11362136 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map