Assay Details
Target Gene Details
Entrez Gene ID: | 285521 |
Gene Name: | COX18, cytochrome c oxidase assembly factor |
Gene Aliases: |
COX18HS |
Location: |
Chr.4:73054696-73069759 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX18 | NM_001297732.1 | 1 | 68 | NP_001284661.1 |
| NM_001297733.1 | 1 | 68 | NP_001284662.1 | |
| NM_001300729.1 | 1 | 68 | NP_001287658.1 | |
| NM_173827.3 | 1 | 68 | NP_776188.1 | |
| XM_005265680.4 | 1 | 68 | XP_005265737.1 | |
| XM_011531878.2 | XP_011530180.1 | |||
| XM_017008045.1 | 1 | 68 | XP_016863534.1 | |
| AK096310.2 | BAC04758.1 | |||
| AY957564.1 | 1 | 64 | AAY35060.1 | |
| AY957565.1 | 1 | 64 | AAY35061.1 | |
| BC071812.1 | 1 | 64 | AAH71812.1 | |
| BC101684.1 | AAI01685.1 | |||
| BC143642.1 | ||||
| DA385228.1 | 1 | 68 | ||
| DA674605.1 | 1 | 68 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523169 | Chr.4:72886525 - 73255193 on Build GRCh38 | Gain |
|
| esv3600985 | Chr.4:72952867 - 73192514 on Build GRCh38 | Gain |
|
| esv3600984 | Chr.4:72944988 - 73087723 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map