Assay Details
Target Gene Details
Entrez Gene ID: | 9472 |
Gene Name: | A-kinase anchoring protein 6 |
Gene Aliases: |
ADAP100, ADAP6, AKAP100, PRKA6, mAKAP |
Location: |
Chr.14:32329273-32833999 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AKAP6 | NM_004274.4 | NP_004265.3 | ||
| AB002309.2 | ||||
| BC137232.1 | ||||
| BC137233.1 | ||||
| BC150185.1 | ||||
| BC150288.1 | ||||
| BC154413.1 | ||||
| BP230135.1 | ||||
| U17195.2 | AAA92354.2 |
More Information
Set Membership: |
|

Genomic Map