Assay Details
Target Gene Details
Entrez Gene ID: | 4054 |
Gene Name: | latent transforming growth factor beta binding protein 3 |
Gene Aliases: |
DASS, LTBP-3, LTBP2, STHAG6, pp6425 |
Location: |
Chr.11:65538559-65558388 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LTBP3 | NM_001130144.2 | 17 | 2702 | NP_001123616.1 |
| NM_001164266.1 | 17 | 2698 | NP_001157738.1 | |
| NM_021070.4 | 17 | 2702 | NP_066548.2 | |
| XM_011545032.2 | 17 | 2884 | XP_011543334.1 | |
| XM_011545033.2 | 17 | 2883 | XP_011543335.1 | |
| AF135960.3 | 17 | 2433 | AAF62352.3 | |
| AF318354.1 | 15 | 1735 | AAL55861.1 | |
| AK024477.1 | 17 | 2699 | ||
| AK090750.1 | 2 | 339 | ||
| AK097212.1 | 2 | 648 | ||
| AK124524.1 | 6 | 1766 | ||
| AK127484.1 | 1 | 2301 | ||
| AK298858.1 | 12 | 1384 | ||
| AK299351.1 | 18 | 2364 | ||
| AL117551.1 | 1 | 57 | CAB55988.1 | |
| BC008761.2 | 8 | 895 | AAH08761.2 | |
| BC136277.1 | 17 | 2508 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951018 | Chr.11:65525430 - 65662029 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map