Assay Details
Target Gene Details
Entrez Gene ID: | 30820 |
Gene Name: | potassium voltage-gated channel interacting protein 1 |
Gene Aliases: |
KCHIP1, VABP |
Location: |
Chr.5:170353055-170736632 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNIP1 | NM_001034838.2 | NP_001030010.1 | ||
| XM_017009407.1 | XP_016864896.1 | |||
| XM_017009408.1 | XP_016864897.1 | |||
| AK301775.1 | ||||
| DC420716.1 | ||||
| DQ148476.1 | AAZ77793.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471054 | Chr.5:170278287 - 170532433 on Build GRCh38 | Loss |
|
| nsv462515 | Chr.5:170265550 - 170532433 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map