Assay Details
Target Gene Details
Entrez Gene ID: | 1891 |
Gene Name: | enoyl-CoA hydratase 1 |
Gene Aliases: |
HPXEL |
Location: |
Chr.19:38815422-38831857 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ECH1 | NM_001398.2 | NP_001389.2 | ||
| BG527600.1 |
More Information
Set Membership: |
|

Genomic Map