Assay Details
Target Gene Details
Entrez Gene ID: | 23522 |
Gene Name: | lysine acetyltransferase 6B |
Gene Aliases: |
GTPTS, MORF, MOZ2, MYST4, ZC2HC6B, qkf, querkopf |
Location: |
Chr.10:74824927-75032623 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KAT6B | NM_001256468.1 | 3 | 814 | NP_001243397.1 |
| NM_001256469.1 | 3 | 814 | NP_001243398.1 | |
| NM_012330.3 | 3 | 814 | NP_036462.2 | |
| XM_005269664.2 | 3 | 875 | XP_005269721.1 | |
| XM_017016000.1 | 3 | 723 | XP_016871489.1 | |
| XM_017016001.1 | 3 | 587 | XP_016871490.1 | |
| XM_017016002.1 | 3 | 826 | XP_016871491.1 | |
| XM_017016003.1 | 2 | 497 | XP_016871492.1 | |
| XM_017016004.1 | 3 | 723 | XP_016871493.1 | |
| XM_017016005.1 | 3 | 723 | XP_016871494.1 | |
| XM_017016006.1 | 3 | 723 | XP_016871495.1 | |
| XM_017016007.1 | 3 | 586 | XP_016871496.1 | |
| XM_017016008.1 | 2 | 497 | XP_016871497.1 | |
| XM_017016009.1 | 3 | 814 | XP_016871498.1 | |
| AB002381.2 | 1 | 113 | BAA20837.2 | |
| AF113514.1 | 2 | 427 | AAF00095.1 | |
| AF119230.1 | 2 | 427 | AAF00099.1 | |
| AF119231.1 | 2 | 427 | AAF00100.1 | |
| AF217500.1 | 3 | 601 | AAL56647.1 | |
| AK074816.1 | 3 | 627 | ||
| BC014143.2 | 3 | 606 | AAH14143.1 | |
| BC048199.1 | 3 | 606 | AAH48199.1 | |
| BC150270.1 | 1 | 140 | ||
| BC150618.1 | 3 | 574 |
More Information
Set Membership: |
|

Genomic Map