Assay Details
Target Gene Details
Entrez Gene ID: | 79625 |
Gene Name: | neuron derived neurotrophic factor |
Gene Aliases: |
C4orf31, NORD |
Location: |
Chr.4:121035627-121072518 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDNF | NM_024574.3 | 4 | 2333 | NP_078850.3 |
| AK026844.1 | 1 | 1418 | BAB15573.1 | |
| AK289615.1 | 4 | 2241 | ||
| AY358821.1 | 4 | 1905 | AAQ89180.1 | |
| BC019351.1 | 4 | 2229 |
More Information
Additional Information:
For this assay, SNP(s) [rs76137074] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map