Assay Details
Target Gene Details
Entrez Gene ID: | 344805 |
Gene Name: | transmembrane protease, serine 7 |
Gene Aliases: |
- |
Location: |
Chr.3:112039067-112081269 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS7 | NM_001042575.2 | NP_001036040.2 | ||
| NR_026734.1 | ||||
| XM_011512754.1 | XP_011511056.1 | |||
| AK131211.1 | BAD18401.1 | |||
| BC117322.1 |
More Information
Set Membership: |
|

Genomic Map