Assay Details
Target Gene Details
Entrez Gene ID: | 57630 |
Gene Name: | SH3 domain containing ring finger 1 |
Gene Aliases: |
POSH, RNF142, SH3MD2 |
Location: |
Chr.4:169094256-169271098 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3RF1 | NM_020870.3 | 10 | 2169 | NP_065921.2 |
| AB040927.1 | 6 | 1084 | BAA96018.1 | |
| AK021429.1 | 10 | 2030 | BAB13822.1 | |
| AK025255.1 | 6 | 1001 | ||
| BC033203.1 | 9 | 1716 | ||
| BC041023.1 | 8 | 1352 | AAH41023.1 | |
| BC053671.1 | 8 | 2511 | AAH53671.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77113673] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map