Assay Details
Target Gene Details
Entrez Gene ID: | 114034 |
Gene Name: | target of EGR1, member 1 (nuclear) |
Gene Aliases: |
hCaf1z |
Location: |
Chr.1:45339457-45343978 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOE1 | NM_025077.3 | 8 | 1589 | NP_079353.3 |
| XM_005270412.3 | 8 | 1572 | XP_005270469.1 | |
| XM_005270413.4 | 8 | 1849 | XP_005270470.1 | |
| XM_011540569.2 | 7 | 1705 | XP_011538871.1 | |
| AK024011.1 | 8 | 1110 | BAB14774.1 | |
| AK093320.1 | 8 | 1384 | ||
| AK293704.1 | 6 | 963 | ||
| AY169960.1 | 8 | 1586 | AAN75441.1 | |
| BC009364.2 | 8 | 1023 | AAH09364.1 | |
| CR457320.1 | 8 | 1006 | CAG33601.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2750837 | Chr.1:45330443 - 45442591 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map