Assay Details
Target Gene Details
Entrez Gene ID: | 102465133 |
Gene Name: | microRNA 6125 |
Gene Aliases: |
hsa-mir-6125, mir-6125 |
Location: |
Chr.12:62260359-62260454 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR6125 | NR_106740.1 |
Target Gene Details
Entrez Gene ID: | 9958 |
Gene Name: | ubiquitin specific peptidase 15 |
Gene Aliases: |
UNPH-2, UNPH4 |
Location: |
Chr.12:62260340-62409721 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP15 | NM_001252078.1 | 1 | 79 | NP_001239007.1 |
| NM_001252079.1 | 1 | 79 | NP_001239008.1 | |
| NM_006313.2 | 1 | 79 | NP_006304.1 | |
| XM_006719718.3 | 1 | 77 | XP_006719781.1 | |
| XM_017020285.1 | 1 | 30 | XP_016875774.1 | |
| AB011101.2 | BAA25455.2 | |||
| AF013990.1 | AAG28973.1 | |||
| AF106069.1 | AAD52099.1 | |||
| AF153604.1 | 1 | 61 | AAD41086.1 | |
| AK023703.1 | BAB14648.1 | |||
| AK292337.1 | 1 | 39 | ||
| AK295947.1 | ||||
| AK297759.1 | ||||
| BC020688.1 | AAH20688.1 | |||
| BC063454.1 | AAH63454.1 | |||
| BC110908.1 | AAI10909.1 | |||
| BC125123.1 | ||||
| DB451322.1 | 1 | 81 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952195 | Chr.12:62256820 - 62286919 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map