Assay Details
Target Gene Details
Entrez Gene ID: | 11073 |
Gene Name: | topoisomerase (DNA) II binding protein 1 |
Gene Aliases: |
TOP2BP1 |
Location: |
Chr.3:133600605-133662380 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOPBP1 | NM_007027.3 | NP_008958.2 | ||
| XM_005247076.3 | XP_005247133.1 | |||
| XM_011512357.2 | XP_011510659.1 | |||
| XM_017005636.1 | XP_016861125.1 | |||
| XM_017005637.1 | XP_016861126.1 | |||
| AB019397.1 | BAA34202.1 | |||
| AK302584.1 | ||||
| BC126209.1 | ||||
| BC151237.1 | ||||
| D87448.1 | ||||
| Y18557.1 |
More Information
Set Membership: |
|

Genomic Map