Assay Details
Target Gene Details
Entrez Gene ID: | 54842 |
Gene Name: | major facilitator superfamily domain containing 6 |
Gene Aliases: |
MMR2, hMMR2 |
Location: |
Chr.2:190408355-190502315 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD6 | NM_017694.3 | 3 | 816 | NP_060164.3 |
| XM_005246654.2 | 2 | 694 | XP_005246711.1 | |
| XM_005246656.3 | 2 | 694 | XP_005246713.1 | |
| XM_011511368.1 | 4 | 882 | XP_011509670.1 | |
| XM_017004392.1 | 3 | 816 | XP_016859881.1 | |
| AB472299.1 | 1 | 492 | ||
| AK098085.1 | 1 | 227 | ||
| AK127176.1 | 3 | 816 | BAC86869.1 |

Genomic Map