Assay Details
Target Gene Details
Entrez Gene ID: | 4734 |
Gene Name: | neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase |
Gene Aliases: |
NEDD4-1, RPF1 |
Location: |
Chr.15:55826917-55993746 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEDD4 | NM_001284338.1 | 15 | 3276 | NP_001271267.1 |
| NM_001284339.1 | 15 | 3228 | NP_001271268.1 | |
| NM_001284340.1 | 14 | 3225 | NP_001271269.1 | |
| NM_006154.3 | 19 | 1910 | NP_006145.2 | |
| NM_198400.3 | 12 | 3060 | NP_940682.2 | |
| NR_104302.1 | 13 | 3163 | ||
| XM_011521624.2 | 18 | 1675 | XP_011519926.1 | |
| XM_011521625.2 | 20 | 1848 | XP_011519927.1 | |
| XM_011521626.1 | 18 | 1692 | XP_011519928.1 | |
| XM_011521627.1 | 16 | 1515 | XP_011519929.1 | |
| AK304394.1 | 19 | 1776 | ||
| AK307541.1 | 15 | 1527 | ||
| AL832063.1 | 12 | 3061 | ||
| AL832359.1 | 14 | 1748 | ||
| AL833293.1 | 14 | 2501 | ||
| AL833300.1 | 15 | 2258 | ||
| AY550969.1 | 19 | 1789 | ||
| BC136605.1 | 15 | 3253 | ||
| BC144284.1 | 15 | 3253 | ||
| BC144285.1 | 15 | 3205 | ||
| BC152452.1 | 19 | 1801 | ||
| BC152562.1 | 19 | 1801 | ||
| D42055.1 | 19 | 1801 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833017 | Chr.15:55673759 - 55865714 on Build GRCh38 | Loss |
|
| nsv569539 | Chr.15:55430015 - 55963576 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map