Assay Details
Target Gene Details
Entrez Gene ID: | 55471 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 7 |
Gene Aliases: |
C2orf56, MidA, PRO1853 |
Location: |
Chr.2:37231631-37259052 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFAF7 | NM_001083946.1 | NP_001077415.1 | ||
| NM_144736.4 | NP_653337.1 | |||
| XM_005264410.1 | XP_005264467.1 | |||
| XM_005264411.1 | XP_005264468.1 | |||
| XM_011532956.1 | XP_011531258.1 | |||
| AK294240.1 | ||||
| AK299015.1 | ||||
| AK302150.1 | ||||
| AK303555.1 | ||||
| BC004548.2 | AAH04548.2 | |||
| BI334629.1 | ||||
| BM172474.1 | ||||
| BX538031.1 | CAD97976.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2684 | Chr.2:37203356 - 37236290 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map