Assay Details
Target Gene Details
Entrez Gene ID: | 6299 |
Gene Name: | spalt like transcription factor 1 |
Gene Aliases: |
HEL-S-89, HSAL1, Sal-1, TBS, ZNF794 |
Location: |
Chr.16:51135975-51152386 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SALL1 | NM_002968.2 | NP_002959.2 | ||
| XM_006721241.3 | XP_006721304.1 | |||
| XM_011523254.2 | XP_011521556.1 | |||
| AK295103.1 | ||||
| BC113881.1 | ||||
| BC113906.1 | ||||
| BC131548.1 | ||||
| BP229682.1 | ||||
| DA318592.1 | ||||
| EU668343.1 | ||||
| Y18265.1 | CAB41400.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2658882 | Chr.16:51150734 - 51156581 on Build GRCh38 | Deletion |
|
| nsv952675 | Chr.16:51150090 - 51157789 on Build GRCh38 | Deletion |
|
| nsv833224 | Chr.16:51062577 - 51219186 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map