Assay Details
Target Gene Details
Entrez Gene ID: | 80332 |
Gene Name: | ADAM metallopeptidase domain 33 |
Gene Aliases: |
C20orf153, DJ964F7.1 |
Location: |
Chr.20:3667973-3682131 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM33 | NM_001282447.2 | 15 | 2342 | NP_001269376.1 |
| NM_025220.4 | 15 | 2342 | NP_079496.1 | |
| NM_153202.3 | 15 | 2342 | NP_694882.1 | |
| XM_005260843.1 | 15 | 1799 | XP_005260900.1 | |
| XM_006723639.1 | 15 | 1799 | XP_006723702.1 | |
| XM_006723640.1 | 15 | 1790 | XP_006723703.1 | |
| XM_011529366.1 | 15 | 1698 | XP_011527668.1 | |
| XM_011529367.1 | 15 | 1659 | XP_011527669.1 | |
| XM_011529368.1 | 15 | 1799 | XP_011527670.1 | |
| XM_011529370.1 | XP_011527672.1 | |||
| XM_011529373.1 | 14 | 1773 | XP_011527675.1 | |
| AB055891.1 | 15 | 1743 | BAB83092.1 | |
| AF466287.1 | 15 | 1661 | AAM80482.1 | |
| AK092627.1 | 1 | 547 | ||
| AK095953.1 | BAC04660.1 | |||
| AK123015.1 | 11 | 2366 | ||
| AK126487.1 | 9 | 3435 | ||
| AK126498.1 | 5 | 2942 | ||
| AK226148.1 | 6 | 3484 | ||
| AL117415.1 | 5 | 1295 | ||
| AY223850.1 | AAP15439.1 | |||
| AY223851.1 | AAP15440.1 | |||
| AY223852.1 | AAP15441.1 | |||
| AY358314.1 | 15 | 1769 | AAQ88680.1 | |
| BC125112.1 | ||||
| BC125113.1 | 13 | 1343 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575355 | Chr.20:3667953 - 3752840 on Build GRCh38 | Gain |
|
| nsv520565 | Chr.20:3652593 - 3740054 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map