Assay Details
Target Gene Details
Entrez Gene ID: | 4151 |
Gene Name: | myoglobin |
Gene Aliases: |
PVALB, myoglobgin |
Location: |
Chr.22:35606764-35623354 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MB | NM_005368.2 | 3 | 485 | NP_005359.1 |
| NM_203377.1 | 4 | 577 | NP_976311.1 | |
| NM_203378.1 | 4 | 560 | NP_976312.1 | |
| XM_005261605.2 | 4 | 521 | XP_005261662.1 | |
| AK300657.1 | 4 | 443 | ||
| AK314057.1 | 4 | 546 | ||
| BC014547.1 | 3 | 481 | AAH14547.1 | |
| BC018001.1 | 3 | 450 | AAH18001.1 | |
| BU585249.1 | ||||
| CB140824.1 | ||||
| CR456516.1 | 3 | 453 | CAG30402.1 | |
| CR541949.1 | 3 | 405 | CAG46747.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510806 | Chr.22:35599609 - 35636220 on Build GRCh38 | Deletion |
|
| nsv834184 | Chr.22:35502044 - 35621639 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map