Assay Details
Target Gene Details
Entrez Gene ID: | 55701 |
Gene Name: | Rho guanine nucleotide exchange factor 40 |
Gene Aliases: |
SOLO |
Location: |
Chr.14:21061276-21090248 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF40 | NM_001278529.1 | 5 | 1841 | NP_001265458.1 |
| NM_001278530.1 | 5 | 1674 | NP_001265459.1 | |
| NM_018071.4 | 5 | 1841 | NP_060541.3 | |
| XM_005267844.2 | 5 | 1877 | XP_005267901.1 | |
| XM_011536937.2 | 5 | 1877 | XP_011535239.1 | |
| XM_011536938.2 | 5 | 1879 | XP_011535240.1 | |
| XM_017021434.1 | 6 | 2134 | XP_016876923.1 | |
| XM_017021435.1 | XP_016876924.1 | |||
| XM_017021436.1 | XP_016876925.1 | |||
| XM_017021437.1 | XP_016876926.1 | |||
| AK024463.1 | 3 | 1079 | ||
| AK074057.1 | 5 | 1786 | ||
| AK126755.1 | ||||
| BC142692.1 | 5 | 1784 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv123n21 | Chr.14:21072704 - 21106531 on Build GRCh38 | Loss |
|
| nsv832740 | Chr.14:20863901 - 21076893 on Build GRCh38 | Gain |
|
| nsv826882 | Chr.14:21042862 - 21136042 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map