Assay Details
Target Gene Details
Entrez Gene ID: | 91582 |
Gene Name: | ribosomal protein S19 binding protein 1 |
Gene Aliases: |
AROS, S19BP |
Location: |
Chr.22:39529093-39532855 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS19BP1 | NM_194326.3 | NP_919307.1 | ||
| NR_130151.1 | ||||
| AK025423.1 | ||||
| AK055865.1 | ||||
| BC037573.1 | AAH37573.1 | |||
| BC047711.1 | AAH47711.1 | |||
| BQ675376.1 | ||||
| CB142920.1 | ||||
| CR456443.1 | CAG30329.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834199 | Chr.22:39324389 - 39531343 on Build GRCh38 | Loss |
|
| nsv3629 | Chr.22:39504067 - 39534583 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116823887] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map