Assay Details
Target Gene Details
Entrez Gene ID: | 9728 |
Gene Name: | SECIS binding protein 2 like |
Gene Aliases: |
SBP2L, SLAN |
Location: |
Chr.15:48988638-49046563 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SECISBP2L | NM_001193489.1 | 18 | 4158 | NP_001180418.1 |
| NM_014701.3 | 17 | 4023 | NP_055516.2 | |
| D87445.3 | 17 | 3893 |
More Information
Set Membership: |
|

Genomic Map