Assay Details
Target Gene Details
Entrez Gene ID: | 27429 |
Gene Name: | HtrA serine peptidase 2 |
Gene Aliases: |
OMI, PARK13, PRSS25 |
Location: |
Chr.2:74529405-74533556 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTRA2 | NM_001321727.1 | 7 | 1972 | NP_001308656.1 |
| NM_001321728.1 | 7 | 1942 | NP_001308657.1 | |
| NM_013247.4 | 8 | 2038 | NP_037379.1 | |
| NM_145074.2 | 6 | 1747 | NP_659540.1 | |
| NR_135769.1 | 8 | 2078 | ||
| NR_135770.1 | 9 | 1506 | ||
| NR_135771.1 | 9 | 1490 | ||
| NR_135772.1 | 9 | 1510 | ||
| AF020760.2 | 8 | 1453 | AAB94569.2 | |
| AF141305.1 | 8 | 2038 | AAF66596.1 | |
| AF141306.1 | 7 | 1331 | AAF66597.1 | |
| AF141307.1 | 7 | 1499 | AAF66598.1 | |
| AF184911.1 | 6 | 1162 | AAG13126.1 | |
| AK225932.1 | 7 | 1375 | ||
| AK291977.1 | ||||
| BC000096.2 | 8 | 1464 | AAH00096.1 |
Target Gene Details
Entrez Gene ID: | 84695 |
Gene Name: | lysyl oxidase like 3 |
Gene Aliases: |
LOXL |
Location: |
Chr.2:74532258-74555709 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOXL3 | NM_001289164.1 | 12 | 2490 | NP_001276093.1 |
| NM_001289165.1 | 10 | 2207 | NP_001276094.1 | |
| NM_032603.3 | 14 | 2925 | NP_115992.1 | |
| AF311313.1 | 14 | 2896 | AAK63205.1 | |
| DQ378059.1 | 10 | 2207 | ABD23013.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759061 | Chr.2:74252797 - 74535298 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map