Assay Details
Target Gene Details
Entrez Gene ID: | 100529261 |
Gene Name: | CHURC1-FNTB readthrough |
Gene Aliases: |
- |
Location: |
Chr.14:64914361-65062655 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHURC1-FNTB | NM_001202558.1 | NP_001189487.1 | ||
| NM_001202559.1 | NP_001189488.1 | |||
| AK296850.1 | ||||
| AK303739.1 |
Target Gene Details
Entrez Gene ID: | 2877 |
Gene Name: | glutathione peroxidase 2 |
Gene Aliases: |
GI-GPx, GPRP, GPRP-2, GPx-2, GPx-GI, GSHPX-GI, GSHPx-2 |
Location: |
Chr.14:64939152-64942905 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPX2 | NM_002083.3 | 2 | 582 | NP_002074.2 |
| NR_046320.1 | 3 | 788 | ||
| NR_046321.1 | 3 | 787 | ||
| AK225871.1 | 3 | 629 | ||
| BC005277.1 | 2 | 422 | AAH05277.1 | |
| BC016756.1 | 2 | 436 | AAH16756.1 | |
| BC022820.1 | 2 | 405 | AAH22820.2 | |
| BC067221.1 | 2 | 426 | AAH67221.1 | |
| BG429974.1 | 2 | 583 | ||
| X53463.1 | 2 | 454 | CAB43534.1 | |
| X68314.1 | 2 | 437 | CAA48394.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2748779 | Chr.14:63663324 - 66595809 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map