Assay Details
Target Gene Details
Entrez Gene ID: | 1656 |
Gene Name: | DEAD-box helicase 6 |
Gene Aliases: |
HLR2, P54, RCK |
Location: |
Chr.11:118747763-118791263 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX6 | NM_001257191.2 | NP_001244120.1 | ||
| NM_004397.5 | NP_004388.2 | |||
| XM_005271417.3 | XP_005271474.1 | |||
| XM_011542644.1 | XP_011540946.1 | |||
| XM_011542645.1 | XP_011540947.1 | |||
| XM_017017251.1 | XP_016872740.1 | |||
| AK093657.1 | ||||
| AK313244.1 | ||||
| BC039826.1 | AAH39826.1 | |||
| BC065007.1 | AAH65007.1 | |||
| D17532.1 | BAA04482.1 | |||
| Z11685.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832283 | Chr.11:118566790 - 118786484 on Build GRCh38 | Loss |
|
| esv2762946 | Chr.11:118714898 - 118762551 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map