Assay Details
Target Gene Details
Entrez Gene ID: | 11282 |
Gene Name: | mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme B |
Gene Aliases: |
GNT-IV, GNT-IVB |
Location: |
Chr.5:179797597-179806952 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGAT4B | NM_014275.4 | NP_055090.1 | ||
| XM_017008998.1 | XP_016864487.1 | |||
| AB000624.1 | BAA83464.1 | |||
| AL537522.3 | ||||
| AY358984.1 | AAQ89343.1 | |||
| BC051835.1 | AAH51835.1 |
Target Gene Details
Entrez Gene ID: | 8878 |
Gene Name: | sequestosome 1 |
Gene Aliases: |
A170, FTDALS3, OSIL, PDB3, ZIP3, p60, p62, p62B |
Location: |
Chr.5:179806388-179838078 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SQSTM1 | NM_001142298.1 | 1 | 147 | NP_001135770.1 |
| BC001874.1 | 1 | 136 | AAH01874.1 | |
| BI549805.1 | 1 | 152 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1133621 | Chr.5:179805800 - 179807200 on Build GRCh38 | Deletion |
|
| nsv469579 | Chr.5:179757830 - 179933191 on Build GRCh38 | Loss |
|
| nsv600668 | Chr.5:179800865 - 179815581 on Build GRCh38 | Loss |
|
| esv3570596 | Chr.5:179804973 - 179813403 on Build GRCh38 | Loss |
|
| esv988291 | Chr.5:179806076 - 179813737 on Build GRCh38 | Deletion |
|
| nsv823361 | Chr.5:179786323 - 179824243 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map