Assay Details
Target Gene Details
Entrez Gene ID: | 83442 |
Gene Name: | SH3 domain binding glutamate rich protein like 3 |
Gene Aliases: |
HEL-S-297, SH3BP-1, TIP-B1 |
Location: |
Chr.1:26279722-26281522 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3BGRL3 | NM_031286.3 | 1 | 344 | NP_112576.1 |
| AF247790.1 | AAL95695.1 | |||
| AF304163.1 | AAG41412.1 | |||
| AF466367.1 | 1 | 892 | AAO33388.1 | |
| AJ297915.1 | CAC35770.1 | |||
| AK024884.1 | ||||
| BC030135.2 | 1 | 113 | AAH30135.1 | |
| DB482618.1 | 1 | 344 | ||
| FJ224343.1 | 1 | 892 |
More Information
Set Membership: |
|

Genomic Map