Assay Details
Target Gene Details
Entrez Gene ID: | 120939 |
Gene Name: | transmembrane protein 52B |
Gene Aliases: |
C12orf59 |
Location: |
Chr.12:10170599-10191804 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM52B | NM_001079815.1 | NP_001073283.1 | ||
| NM_153022.3 | 1 | 85 | NP_694567.1 | |
| XM_006719029.2 | 1 | 85 | XP_006719092.1 | |
| AK055728.1 | BAB70998.1 | |||
| AY358845.1 | AAQ89204.1 | |||
| DA156941.1 | ||||
| DA621425.1 | 1 | 85 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3628526 | Chr.12:10143326 - 10179529 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map