Assay Details
Target Gene Details
Entrez Gene ID: | 221476 |
Gene Name: | peptidase inhibitor 16 |
Gene Aliases: |
CD364, CRISP9, MSMBBP, PSPBP |
Location: |
Chr.6:36948263-36964837 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PI16 | NM_001199159.1 | 6 | 1179 | NP_001186088.1 |
| NM_153370.2 | 5 | 1284 | NP_699201.2 | |
| XM_005248917.2 | XP_005248974.1 | |||
| XM_011514375.2 | 5 | 1113 | XP_011512677.1 | |
| XM_017010430.1 | XP_016865919.1 | |||
| AK075470.1 | 5 | 979 | BAC11640.1 | |
| AK124589.1 | BAC85892.1 | |||
| AK302193.1 | 4 | 787 | ||
| AK303928.1 | 4 | 933 | ||
| AK312785.1 | 5 | 977 | ||
| AY358422.1 | 5 | 970 | AAQ88788.1 | |
| BC022399.1 | 5 | 978 | AAH22399.2 | |
| BC035634.1 | 5 | 1284 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map