Assay Details
Target Gene Details
Entrez Gene ID: | 84971 |
Gene Name: | autophagy related 4D cysteine peptidase |
Gene Aliases: |
APG4-D, APG4D, AUTL4 |
Location: |
Chr.19:10543894-10553423 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATG4D | NM_001281504.1 | NP_001268433.1 | ||
| NM_032885.5 | NP_116274.3 | |||
| NR_104024.1 | ||||
| NR_104025.1 | ||||
| XM_006722927.1 | XP_006722990.1 | |||
| AK056210.1 | BAB71121.1 | |||
| BC007639.1 | AAH07639.1 | |||
| BC016845.1 | AAH16845.1 | |||
| BC068992.1 | AAH68992.1 | |||
| BM671285.1 |
Target Gene Details
Entrez Gene ID: | 65095 |
Gene Name: | KRI1 homolog |
Gene Aliases: |
- |
Location: |
Chr.19:10553085-10566031 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRI1 | NM_023008.3 | 19 | 2670 | NP_075384.3 |
| XM_011528190.2 | 18 | 2601 | XP_011526492.1 | |
| AK097181.1 | 10 | 1988 | ||
| AL833189.1 | 17 | 3409 | ||
| BM676050.1 | 1 | 354 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833744 | Chr.19:10458964 - 10635033 on Build GRCh38 | Loss |
|
| nsv833745 | Chr.19:10539132 - 10727920 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map