Assay Details
Target Gene Details
Entrez Gene ID: | 64864 |
Gene Name: | regulatory factor X7 |
Gene Aliases: |
RFXDC2 |
Location: |
Chr.15:56090533-56247654 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFX7 | NM_022841.5 | 9 | 3397 | NP_073752.5 |
| XM_005254603.3 | 10 | 3491 | XP_005254660.2 | |
| XM_011521925.2 | 9 | 3329 | XP_011520227.1 | |
| XM_017022506.1 | 10 | 6214 | XP_016877995.1 | |
| XM_017022507.1 | 9 | 3457 | XP_016877996.1 | |
| XM_017022508.1 | 8 | 4626 | XP_016877997.1 | |
| AK023056.1 | 1 | 1692 | BAB14381.1 | |
| AK126103.1 | 2 | 2549 | BAC86441.1 | |
| AK128045.1 | 9 | 3334 | BAC87248.1 | |
| AL832584.1 | 1 | 2206 | ||
| AL833367.1 | 1 | 1624 | ||
| AL834302.1 | 1 | 206 | CAD38972.1 | |
| BC112936.1 | 7 | 3217 | AAI12937.1 | |
| BC143438.1 | 7 | 3154 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761872 | Chr.15:55918867 - 56134303 on Build GRCh38 | Gain |
|
| nsv833018 | Chr.15:56033601 - 56200337 on Build GRCh38 | Gain |
|
| esv3569620 | Chr.15:55918120 - 56117670 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map