Assay Details
Target Gene Details
Entrez Gene ID: | 116224 |
Gene Name: | family with sequence similarity 122A |
Gene Aliases: |
C9orf42 |
Location: |
Chr.9:68780006-68783879 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM122A | NM_138333.4 | 1 | 286 | NP_612206.4 |
| AK126379.1 | 1 | 227 | ||
| BC013062.1 | 1 | 237 | AAH13062.1 | |
| BI757526.1 | 1 | 244 | ||
| HY099611.1 | 1 | 286 |
Target Gene Details
Entrez Gene ID: | 8395 |
Gene Name: | phosphatidylinositol-4-phosphate 5-kinase type 1 beta |
Gene Aliases: |
MSS4, STM7 |
Location: |
Chr.9:68705217-69009176 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIP5K1B | NM_001278253.1 | NP_001265182.1 | ||
| NM_003558.3 | NP_003549.1 | |||
| XM_005252261.3 | XP_005252318.1 | |||
| XM_005252262.4 | XP_005252319.1 | |||
| XM_006717300.2 | XP_006717363.1 | |||
| XM_006717301.1 | XP_006717364.1 | |||
| XM_011519082.2 | XP_011517384.1 | |||
| XM_011519083.2 | XP_011517385.1 | |||
| XM_011519084.2 | XP_011517386.1 | |||
| XM_017015188.1 | XP_016870677.1 | |||
| XM_017015189.1 | XP_016870678.1 | |||
| XM_017015190.1 | XP_016870679.1 | |||
| XM_017015191.1 | XP_016870680.1 | |||
| AK292734.1 | ||||
| AK295587.1 | ||||
| BC030587.2 | AAH30587.1 | |||
| CB988216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422258 | Chr.9:68619929 - 69048424 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map