Assay Details
Target Gene Details
Entrez Gene ID: | 84126 |
Gene Name: | ATR interacting protein |
Gene Aliases: |
- |
Location: |
Chr.3:48446710-48465655 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATRIP | NM_001271022.1 | 9 | 1204 | NP_001257951.1 |
| NM_001271023.1 | 8 | 1461 | NP_001257952.1 | |
| NM_032166.3 | 8 | 1414 | NP_115542.2 | |
| NM_130384.2 | 8 | 1414 | NP_569055.1 | |
| AF451323.1 | 8 | 1310 | AAL38042.1 | |
| AK022405.1 | 7 | 1009 | BAB14029.1 | |
| AK291568.1 | 6 | 880 | ||
| AK291829.1 | 8 | 1345 | ||
| AK300548.1 | 9 | 1177 | ||
| AK315075.1 | 8 | 1378 | ||
| AL832917.1 | 6 | 879 | CAH10621.1 | |
| BC014153.2 | 8 | 1329 | AAH14153.2 | |
| BC020563.2 | 8 | 1337 | AAH20563.1 | |
| BC030597.2 | 8 | 1461 | AAH30597.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834684 | Chr.3:48344209 - 48551890 on Build GRCh38 | Gain |
|
| nsv590242 | Chr.3:48392157 - 48518906 on Build GRCh38 | Loss |
|
| nsv508917 | Chr.3:48400224 - 48499357 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map