Assay Details
Target Gene Details
Entrez Gene ID: | 10478 |
Gene Name: | solute carrier family 25 member 17 |
Gene Aliases: |
PMP34 |
Location: |
Chr.22:40769630-40819399 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A17 | NM_001282726.1 | 10 | 1844 | NP_001269655.1 |
| NM_001282727.1 | 7 | 1572 | NP_001269656.1 | |
| NM_006358.3 | 9 | 1791 | NP_006349.1 | |
| NR_104235.1 | 11 | 2103 | ||
| NR_104236.1 | 8 | 1639 | ||
| NR_104237.1 | 8 | 1681 | ||
| NR_104238.1 | 6 | 1511 | ||
| AK094062.1 | ||||
| BC005957.1 | AAH05957.1 | |||
| BC012998.2 | AAH12998.1 | |||
| BU686766.1 | 1 | 500 | ||
| BX647991.1 | 8 | 1571 | ||
| Y12860.1 | CAA73367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834205 | Chr.22:40593419 - 40777442 on Build GRCh38 | Gain+Loss |
|
| nsv834206 | Chr.22:40696722 - 40838881 on Build GRCh38 | Gain |
|
| esv3568414 | Chr.22:40739711 - 40771039 on Build GRCh38 | Loss |
|
| nsv1062438 | Chr.22:40646389 - 40806134 on Build GRCh38 | Gain |
|
| nsv1066802 | Chr.22:40725180 - 40776833 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map