Assay Details
Target Gene Details
Entrez Gene ID: | 55970 |
Gene Name: | G protein subunit gamma 12 |
Gene Aliases: |
- |
Location: |
Chr.1:67701466-67833779 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNG12 | NM_018841.5 | 4 | 1405 | NP_061329.3 |
| XM_017001809.1 | 4 | 1572 | XP_016857298.1 | |
| XM_017001810.1 | 5 | 1813 | XP_016857299.1 | |
| XM_017001811.1 | 5 | 1742 | XP_016857300.1 | |
| AK092014.1 | 1 | 594 | ||
| AL832431.1 | 4 | 1358 | CAH10645.1 | |
| BC005940.1 | 4 | 1386 | AAH05940.1 | |
| BU151184.1 | 1 | 197 |
More Information
Additional Information:
For this assay, SNP(s) [rs115148897] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map