Assay Details
Target Gene Details
Entrez Gene ID: | 9878 |
Gene Name: | TOX high mobility group box family member 4 |
Gene Aliases: |
C14orf92, KIAA0737, LCP1, MIG7 |
Location: |
Chr.14:21477176-21499177 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOX4 | NM_001303523.1 | 8 | 2489 | NP_001290452.1 |
| NM_014828.3 | 9 | 2558 | NP_055643.1 | |
| AB018280.1 | 9 | 2487 | ||
| AY305872.1 | 1 | 364 | AAQ95214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3633718 | Chr.14:21495207 - 21527525 on Build GRCh38 | Gain |
|
| esv2422459 | Chr.14:21463465 - 21701280 on Build GRCh38 | Duplication |
|
| esv3633716 | Chr.14:21455908 - 21564274 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map