Assay Details
Target Gene Details
Entrez Gene ID: | 51185 |
Gene Name: | cereblon |
Gene Aliases: |
MRT2, MRT2A |
Location: |
Chr.3:3148490-3179717 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRBN | NM_001173482.1 | 10 | 1127 | NP_001166953.1 |
| NM_016302.3 | 10 | 1130 | NP_057386.2 | |
| XM_005265202.3 | 9 | 1011 | XP_005265259.1 | |
| XM_011533791.2 | XP_011532093.1 | |||
| XM_011533793.2 | 11 | 1258 | XP_011532095.1 | |
| XM_011533794.2 | 10 | 1129 | XP_011532096.1 | |
| AF117230.1 | 10 | 1094 | AAF17211.1 | |
| AK027507.1 | 10 | 1108 | BAB55162.1 | |
| AK058191.1 | 10 | 1104 | ||
| AK123077.1 | 2 | 1503 | ||
| AK301423.1 | 9 | 1002 | ||
| AK312577.1 | ||||
| BC017419.1 | 10 | 1118 | AAH17419.1 | |
| BC067811.1 | 11 | 1111 | AAH67811.1 | |
| BX647351.1 | 2 | 3442 | ||
| CR627060.1 | 6 | 3523 | CAH10361.1 |
Target Gene Details
Entrez Gene ID: | 51095 |
Gene Name: | tRNA nucleotidyl transferase 1 |
Gene Aliases: |
CCA1, CGI-47, MtCCA, RPEM, SIFD |
Location: |
Chr.3:3126916-3153435 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRNT1 | XM_011533778.2 | 9 | 3405 | XP_011532080.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv589371 | Chr.3:2170634 - 3594980 on Build GRCh38 | Loss |
|
| esv2422274 | Chr.3:2100396 - 3314907 on Build GRCh38 | Duplication |
|
| esv2751994 | Chr.3:2200757 - 3178886 on Build GRCh38 | Gain |
|
| dgv4636n100 | Chr.3:2825734 - 3271845 on Build GRCh38 | Gain |
|
| nsv834604 | Chr.3:3147083 - 3361983 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map